Merck
  • Inducible and reversible inhibition of miRNA-mediated gene repression in vivo.
Inducible and reversible inhibition of miRNA-mediated gene repression in vivo.
eLife (2021-09-01)
Gaspare La Rocca, Bryan King, Bing Shui, Xiaoyi Li, Minsi Zhang, Kemal M Akat, Paul Ogrodowski, Chiara Mastroleo, Kevin Chen, Vincenzo Cavalieri, Yilun Ma, Viviana Anelli, Doron Betel, Joana Vidigal, Thomas Tuschl, Gunter Meister, Craig B Thompson, Tullia Lindsten, Kevin Haigis, Andrea Ventura
ABSTRACT
Although virtually all gene networks are predicted to be controlled by miRNAs, the contribution of this important layer of gene regulation to tissue homeostasis in adult animals remains unclear. Gain and loss-of-function experiments have provided key insights into the specific function of individual miRNAs, but effective genetic tools to study the functional consequences of global inhibition of miRNA activity in vivo are lacking. Here we report the generation and characterization of a genetically engineered mouse strain in which miRNA-mediated gene repression can be reversibly inhibited without affecting miRNA biogenesis or abundance. We demonstrate the usefulness of this strategy by investigating the consequences of acute inhibition of miRNA function in adult animals. We find that different tissues and organs respond differently to global loss of miRNA function. While miRNA-mediated gene repression is essential for the homeostasis of the heart and the skeletal muscle, it is largely dispensable in the majority of other organs. Even in tissues where it is not required for homeostasis, such as the intestine and hematopoietic system, miRNA activity can become essential during regeneration following acute injury. These data support a model where many metazoan tissues primarily rely on miRNA function to respond to potentially pathogenic events.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Supelco
2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic acid, pharmaceutical impurity standard
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
ESGRO® Recombinant Mouse LIF Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^7 units/ml.
Sigma-Aldrich
Anti-pan Ago Antibody, clone 2A8, clone 2A8, from mouse