Merck
  • Effect of therapeutic hypothermia against renal injury in a rat model of asphyxial cardiac arrest: Α focus on the survival rate, pathophysiology and antioxidant enzymes.
Effect of therapeutic hypothermia against renal injury in a rat model of asphyxial cardiac arrest: Α focus on the survival rate, pathophysiology and antioxidant enzymes.
Molecular medicine reports (2021-11-20)
So Eun Kim, Ha-Young Shin, Eui-Yong Lee, Yeo-Jin Yoo, Ryun-Hee Kim, Jeong-Hwi Cho, Tae-Kyeong Lee, Dongchoon Ahn, Byung-Yong Park, Jae Chol Yoon, Seongkweon Hong, In-Shik Kim, Hyun-Jin Tae, Moo-Ho Won
ABSTRACT
Although multi‑organ dysfunction is associated with the survival rate following cardiac arrest (CA), the majority of studies to date have focused on hearts and brains, and few studies have considered renal failure. The objective of the present study, therefore, was to examine the effects of therapeutic hypothermia on the survival rate, pathophysiology and antioxidant enzymes in rat kidneys following asphyxial CA. Rats were sacrificed one day following CA. The survival rate, which was estimated using Kaplan‑Meier analysis, was 42.9% one day following CA. However, hypothermia, which was induced following CA, significantly increased the survival rate (71.4%). In normothermia rats with CA, the serum blood urea nitrogen level was significantly increased one day post‑CA. In addition, the serum creatinine level was significantly increased one day post‑CA. However, in CA rats exposed to hypothermia, the levels of urea nitrogen and creatinine significantly decreased following CA. Histochemical staining revealed a significant temporal increase in renal injury after the normothermia group was subjected to CA. However, renal injury was significantly decreased in the hypothermia group. Immunohistochemical analysis of the kidney revealed a significant decrease in antioxidant enzymes (copper‑zinc superoxide dismutase, manganese superoxide dismutase, glutathione peroxidase and catalase) with time in the normothermia group. However, in the hypothermia group, these enzymes were significantly elevated following CA. Collectively, the results revealed that renal dysfunction following asphyxial CA was strongly associated with the early survival rate and therapeutic hypothermia reduced renal injury via effective antioxidant mechanisms.
Sigma-Aldrich
Anti-SOD1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-SOD2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution