Merck
  • Protocol for in vivo and ex vivo assessments of glucose-stimulated insulin secretion in mouse islet β cells.
Protocol for in vivo and ex vivo assessments of glucose-stimulated insulin secretion in mouse islet β cells.
STAR protocols (2021-08-20)
Yun-Xia Zhu, Yun-Cai Zhou, Yan Zhang, Peng Sun, Xiao-Ai Chang, Xiao Han
ABSTRACT
Pancreatic islet β cells secrete insulin in a biphasic manner when sensing high blood glucose level. This protocol describes the evaluation of different phases of insulin secretion, as well as basal, glucose-stimulated and total insulin secretion abilities, thereby enabling precise assessment of β cell function both in vivo and ex vivo. The in vivo assay consists of intravenous tube imbedding surgery and hyperglycemic clamp. The ex vivo assay consists of islet isolation, dynamic perfusion and static immersion. For complete details on the use and execution of this protocol, please refer to Sun et al. (2021).
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type V, ≥1 FALGPA units/mg solid, >125 CDU/mg solid
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077 g/mL
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, ACS reagent, ≥99.0%
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Magnesium chloride hexahydrate, ACS reagent, 99.0-102.0%
Sigma-Aldrich
Dithizone, Practical Grade