Merck
Prophages encode phage-defense systems with cognate self-immunity.
Cell host & microbe (2021-10-02)
Siân V Owen, Nicolas Wenner, Charles L Dulberger, Ella V Rodwell, Arthur Bowers-Barnard, Natalia Quinones-Olvera, Daniel J Rigden, Eric J Rubin, Ethan C Garner, Michael Baym, Jay C D Hinton
ABSTRACT
Temperate phages are pervasive in bacterial genomes, existing as vertically inherited islands termed prophages. Prophages are vulnerable to predation of their host bacterium by exogenous phages. Here, we identify BstA, a family of prophage-encoded phage-defense proteins in diverse Gram-negative bacteria. BstA localizes to sites of exogenous phage DNA replication and mediates abortive infection, suppressing the competing phage epidemic. During lytic replication, the BstA-encoding prophage is not itself inhibited by BstA due to self-immunity conferred by the anti-BstA (aba) element, a short stretch of DNA within the bstA locus. Inhibition of phage replication by distinct BstA proteins from Salmonella, Klebsiella, and Escherichia prophages is generally interchangeable, but each possesses a cognate aba element. The specificity of the aba element ensures that immunity is exclusive to the replicating prophage, preventing exploitation by variant BstA-encoding phages. The BstA protein allows prophages to defend host cells against exogenous phage attack without sacrificing the ability to replicate lytically.
Sigma-Aldrich
m-Toluic acid, ReagentPlus®, 99%
Sigma-Aldrich
Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
D-(+)-Maltose monohydrate, from potato, ≥99% (HPLC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Roche
Blocking Reagent, For nucleic acid hybridization and detection
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
M9, Minimal Salts, 5X, powder, minimal microbial growth medium
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Supelco
Anhydrotetracycline hydrochloride, VETRANAL®, analytical standard
Roche
DIG Easy Hyb Granules, powder, suitable for hybridization