Merck
  • Periorbital Nociception in a Progressive Multiple Sclerosis Mouse Model Is Dependent on TRPA1 Channel Activation.
Periorbital Nociception in a Progressive Multiple Sclerosis Mouse Model Is Dependent on TRPA1 Channel Activation.
Pharmaceuticals (Basel, Switzerland) (2021-08-29)
Diéssica Padilha Dalenogare, Camila Ritter, Fernando Roberto Antunes Bellinaso, Sabrina Qader Kudsi, Gabriele Cheiran Pereira, Maria Fernanda Pessano Fialho, Débora Denardin Lückemeyer, Caren Tatiane de David Antoniazzi, Lorenzo Landini, Juliano Ferreira, Guilherme Vargas Bochi, Sara Marchesan Oliveira, Francesco De Logu, Romina Nassini, Pierangelo Geppetti, Gabriela Trevisan
ABSTRACT
Headaches are frequently described in progressive multiple sclerosis (PMS) patients, but their mechanism remains unknown. Transient receptor potential ankyrin 1 (TRPA1) was involved in neuropathic nociception in a model of PMS induced by experimental autoimmune encephalomyelitis (PMS-EAE), and TRPA1 activation causes periorbital and facial nociception. Thus, our purpose was to observe the development of periorbital mechanical allodynia (PMA) in a PMS-EAE model and evaluate the role of TRPA1 in periorbital nociception. Female PMS-EAE mice elicited PMA from day 7 to 14 days after induction. The antimigraine agents olcegepant and sumatriptan were able to reduce PMA. The PMA was diminished by the TRPA1 antagonists HC-030031, A-967079, metamizole and propyphenazone and was absent in TRPA1-deficient mice. Enhanced levels of TRPA1 endogenous agonists and NADPH oxidase activity were detected in the trigeminal ganglion of PMS-EAE mice. The administration of the anti-oxidants apocynin (an NADPH oxidase inhibitor) or alpha-lipoic acid (a sequestrant of reactive oxygen species), resulted in PMA reduction. These results suggest that generation of TRPA1 endogenous agonists in the PMS-EAE mouse model may sensitise TRPA1 in trigeminal nociceptors to elicit PMA. Thus, this ion channel could be a potential therapeutic target for the treatment of headache in PMS patients.
Sigma-Aldrich
Pertussis toxin from Bordetella pertussis, lyophilized powder
Sigma-Aldrich
Freund′s Adjuvant, Incomplete, liquid
Sigma-Aldrich
Acetovanillone, ≥98%, FG
Sigma-Aldrich
A-967079, ≥98% (HPLC)
Sigma-Aldrich
BIBN4096BS, ≥95% (HPLC)
Roche
Tris hydrochloride, >99% (titration), pH 7.0-9.0, suitable for FISH
Sigma-Aldrich
Peroxidase from horseradish, Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
(±)-α-Lipoic acid, synthetic, ≥99% (titration), powder
Sigma-Aldrich
TWEEN® 80, viscous liquid
Sigma-Aldrich
HC-030031, ≥98% (HPLC), powder
Sigma-Aldrich
Cytochrome c Reductase (NADPH) Assay Kit, 1 kit sufficient for 100 tests, determining cytochrome c reductase activity
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Metamizole sodium hydrate, ≥98% (HPLC)