Merck
  • Molecular motor protein KIF5C mediates structural plasticity and long-term memory by constraining local translation.
Molecular motor protein KIF5C mediates structural plasticity and long-term memory by constraining local translation.
Cell reports (2021-07-15)
Supriya Swarnkar, Yosef Avchalumov, Isabel Espadas, Eddie Grinman, Xin-An Liu, Bindu L Raveendra, Aya Zucca, Sonia Mediouni, Abhishek Sadhu, Susana Valente, Damon Page, Kyle Miller, Sathyanarayanan V Puthanveettil
ABSTRACT
Synaptic structural plasticity, key to long-term memory storage, requires translation of localized RNAs delivered by long-distance transport from the neuronal cell body. Mechanisms and regulation of this system remain elusive. Here, we explore the roles of KIF5C and KIF3A, two members of kinesin superfamily of molecular motors (Kifs), and find that loss of function of either kinesin decreases dendritic arborization and spine density whereas gain of function of KIF5C enhances it. KIF5C function is a rate-determining component of local translation and is associated with ∼650 RNAs, including EIF3G, a regulator of translation initiation, and plasticity-associated RNAs. Loss of function of KIF5C in dorsal hippocampal CA1 neurons constrains both spatial and contextual fear memory, whereas gain of function specifically enhances spatial memory and extinction of contextual fear. KIF5C-mediated long-distance transport of local translation substrates proves a key mechanism underlying structural plasticity and memory.
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
1(S),9(R)-(−)-Bicuculline methiodide, ≥95.0% (HPCE)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Anti-Glutamate Receptor 2 Antibody, extracellular, clone 6C4, clone 6C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Kinesin Antibody, heavy chain, a.a.420-445, clone H2, clone H2, Chemicon®, from mouse
Roche
DIG RNA Labeling Kit (SP6/T7), sufficient for 2 x 10 labeling reactions, kit of 1 (12 components), suitable for hybridization, suitable for Southern blotting
Roche
Biotin RNA Labeling Mix, sufficient for 20 reactions (transcription), pkg of 40 μL, solution
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Anti-Puromycin Antibody, clone 4G11, clone 4G11, from mouse
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Anti-GluR2 Antibody, clone L21/32, clone L21/32, from mouse