Merck
  • An optimized protocol with a stepwise approach to identify specific nuclear receptor ligands from cultured mammalian cells.
An optimized protocol with a stepwise approach to identify specific nuclear receptor ligands from cultured mammalian cells.
STAR protocols (2021-07-22)
Alexandre Berthier, Bart Staels, Philippe Lefebvre
ABSTRACT
Here, we describe an optimized protocol to identify specific nuclear receptor ligands. First, to rule out any compound interference with luciferase activity per se, we describe an in vitro assay assessing potential inhibition or activation of luciferase enzymatic activity. Second, to comply with EMA and FDA guidelines to mitigate drug-drug interactions, we detail assays assessing constitutive androstane receptor (CAR) and pregnane X receptor (PXR) activation ability. Finally, to minimize off-target detection effects, we describe the use of mammalian one- (or two-) hybrid systems. For complete details on the use and execution of this protocol, please refer to Hering et al. (2018).
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
PK 11195
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
CITCO, ≥98% (HPLC), solid