Merck
  • Use of L-leucyl-3-carboxy-4-hydroxyanilide as substrate for determining the activity of microsomal aminopeptidase in serum.
Use of L-leucyl-3-carboxy-4-hydroxyanilide as substrate for determining the activity of microsomal aminopeptidase in serum.
Clinical chemistry (1985-10-01)
M Shimamoto, S Takewaki, S Sakuraoka, T Nagasawa, K Kuroiwa, O Kodama, T Akatsuka
ABSTRACT
We describe a colorimetric method for assay of microsomal aminopeptidase (EC 3.4.11.2) activity in serum. We use a new substrate, L-leucyl-3-carboxy-4-hydroxyanilide, p-xylenol as coupler, and sodium metaperiodate as oxidizing reagent. The colored substance formed by the oxidative condensation between p-xylenol and 5-aminosalicylic acid absorbs maximally at 635 nm, and can be directly measured in serum. In a previous method for this enzyme, L-leucyl-beta-naphthylamide was used as substrate and beta-naphthylamine, a carcinogenic reagent used as a standard in making the assay, was unsuitable for routine use. We found a close correlation between results obtained with the new, safer method and with the previous method.
Sigma-Aldrich
3,5-Dimethylphenol, ≥99%
Sigma-Aldrich
3,5-Dimethylphenol, 98%