Merck
  • Metabolites of Prickly Rose: Chemodiversity and Digestive-Enzyme-Inhibiting Potential of Rosa acicularis and the Main Ellagitannin Rugosin D.
Metabolites of Prickly Rose: Chemodiversity and Digestive-Enzyme-Inhibiting Potential of Rosa acicularis and the Main Ellagitannin Rugosin D.
Plants (Basel, Switzerland) (2021-11-28)
Daniil N Olennikov, Vladimir V Chemposov, Nadezhda K Chirikova
ABSTRACT
Prickly rose (Rosaacicularis Lindl.) is the most distributed rose species in the Northern Hemisphere, used by indigenous people for various food purposes. The lack of detailed information about the chemical composition of R. acicularis has led us to study the phytochemical composition and metabolic profile of prickly rose extracts using chromatographic techniques. Many groups of phenolic and non-phenolic compounds were quantified in the leaves, flowers, roots and fruits of R. acicularis. Phenolic compounds were the dominant phytochemicals in the aerial parts and roots of R. acicularis. A precise study by high-performance liquid chromatography with photodiode array detection and electrospray ionization triple quadrupole mass spectrometric detection showed the presence of 123 compounds, among which ellagic acid derivatives, ellagitannins, gallotannins, catechins, catechin oligomers, hydroxycinnamates and flavonoid glycosides of kaempferol, quercetin and dihydroquercetin were all identified for the first time. The most abundant phenolic compounds were ellagitannins and flavonoid glycosides, with a maximal content of 70.04 mg/g in leaves and 66.72 mg/g in flowers, respectively, indicating the great ability of R. acicularis organs to accumulate phenolic compounds. By applying a standardized static, simulated gastrointestinal digestion method, we found the inhibitory potential of the leaf extract against digestive α-amylases. A pancreatic α-amylase activity-inhibiting assay coupled with HPLC microfractionation demonstrated high inhibition of enzyme activity by ellagitannin rugosin D, which was later confirmed by a microplate reaction with mammalian α-amylases and the simulated digestion method. This study clearly demonstrates that R. acicularis leaf extract and its main component, ellagitannin rugosin D, strongly inhibit digestive α-amylase, and may be a prospective antidiabetic agent.
Sigma-Aldrich
Nα-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride, protease substrate
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Colipase from porcine pancreas, essentially salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein