Merck
  • PARP-1-regulated TNF-α expression in the dorsal root ganglia and spinal dorsal horn contributes to the pathogenesis of neuropathic pain in rats.
PARP-1-regulated TNF-α expression in the dorsal root ganglia and spinal dorsal horn contributes to the pathogenesis of neuropathic pain in rats.
Brain, behavior, and immunity (2020-04-14)
Yan Gao, Liying Bai, Wenjuan Zhou, Yin Yang, Jian Zhang, Liren Li, Mingjun Jiang, Yang Mi, Tong-Tong Li, Xuan Zhang, Wei Zhang, Ji-Tian Xu
ABSTRACT
Emerging evidence has implicated poly-(ADP-ribose) polymerase 1 (PARP-1), a transcriptional coregulator, in a variety of inflammatory diseases. In the current study, the role of PARP-1 in neuropathic pain and the underlying mechanisms were investigated. Neuropathic pain was determined by assessing the paw withdrawal threshold (PWT) and paw withdrawal latency (PWL) following lumbar 5 spinal nerve ligation (SNL) in male rates. Western blotting, qRT-PCR, immunohistochemistry, chromatin immunoprecipitation (ChIP), and Co-IP assays were performed to elucidate the mechanisms. The results showed that SNL resulted in a significant increase in the expression and activation of PARP-1 in the ipsilateral L4/5 dorsal root ganglia (DRG) and spinal dorsal horn, which occurred on day one, reached peak on day 7, and persisted more than 2 weeks after surgery. Double immunofluorescence staining revealed that PARP-1 was expressed exclusively in DRG A-type and C-type neurons. In the spinal cord, PARP-1 mainly colocalized with the neuronal marker NeuN and the astrocytic marker GFAP specifically in the superficial lamina. Prior intrathecal (i.t.) injection of PJ-34, a PARPs inhibitor, or Tiq-A, a specific PARP-1 inhibitor, dose-dependently prevented the reductions in PWT and PWL following SNL. Established neuropathic pain-like hypersensitivity was also attenuated with i.t. injection of PJ-34 and Tiq-A starting on day 7 following SNL, a timepoint at which neuropathic pain was fully established. SNL-induced mechanical allodynia and thermal hyperalgesia were also alleviated by i.t. injection of PARP-1 siRNA following a reduction in PARP-1 expression in the dorsal horn. Moreover, the SNL-induced increases in TNF-α protein and mRNA in the dorsal horn and DRG were dramatically suppressed by i.t. injection of Tiq-A or PARP-1 siRNA. The i.t. lipopolysaccharide (LPS)-induced increase in the production of TNF-α in the dorsal horn was also inhibited by prior to i.t. injection of PARP-1 siRNA. Results of ChIP assay showed that SNL-induced PARP-1 activation promoted the binding of NF-κB p65 with the TNF-α promoter in the dorsal horn and that PARP-1 inhibition reduced this binding and suppressed TNF-α expression. Co-IP assay revealed that SNL caused a significant increase in the level of histone H1 poly(ADP)-ribosylation. Together, these results indicate that PARP-1-regulated TNF-α expression in the DRG and spinal dorsal horn following SNL contributes to the development and maintenance of neuropathic pain. Targeting PARP-1 might be a promising therapeutic strategy for the treatment of the chronic pain.
Sigma-Aldrich
PJ-34 hydrochloride hydrate, ≥98% (HPLC), powder