Merck
  • Spoilage indicator bacteria in farmed Atlantic salmon (Salmo salar) stored on ice for 10 days.
Spoilage indicator bacteria in farmed Atlantic salmon (Salmo salar) stored on ice for 10 days.
Food microbiology (2018-10-10)
Colin Fogarty, Paul Whyte, Nigel Brunton, James Lyng, Conor Smyth, John Fagan, Declan Bolton
ABSTRACT
This study investigated the growth of indicator and spoilage bacteria on whole Atlantic salmon (Salmo salar) stored aerobically at 2 °C. On days 0, 2, 3, 6, 8 and 10 microbiological analysis was carried out on inner flesh and outer skin samples as well as outer skin swabs (25 cm2 surface areas). Mesophilic total viable counts (TVCm) on skin, flesh and swab samples increased from 1.9, 1.1 and 2.7 log10 CFUcm2 to 6.0, 5.1 and 5.7 log10 CFU/cm2 after 10 days, respectively. Psychrotrophic counts (TVCp), increased from 2.2, 1.8 and 3.1 log10 CFU/cm2 to 6.2, 5.3 and 5.9 log10 CFU/cm2, for skin, flesh and swab samples respectively. Hydrogen sulphide producing bacteria (HSPB), lactic acid bacteria (LAB), Pseudomonas spp., Brochothrix thermosphacta and Photobacterium spp. grew well with similar growth rates (mean generation times of 17.2-26 h). It was concluded that the shelf-life of salmon at 2 °C was approximately 10 days and that HSPB, LAB, Pseudomonas spp., Br. thermosphacta and Photobacterium spp. may be a better indicator of fish spoilage rather than TVC growth, with a count of 5-6 log10 CFU/cm2 indicating the end of shelf-life.
Product Description
Millipore
Photobacterium Broth, suitable for microbiology, NutriSelect® Plus