Merck
  • Evaluation of vitamin D3 A-ring analogues as Hedgehog pathway inhibitors.
Evaluation of vitamin D3 A-ring analogues as Hedgehog pathway inhibitors.
Bioorganic & medicinal chemistry letters (2012-01-10)
Upasana Banerjee, Manuka Ghosh, M Kyle Hadden
ABSTRACT
A structure-activity relationship study focusing on the A-ring of vitamin D3 (VD3) was undertaken to elucidate its role in inhibiting the Hedgehog pathway and in mediating anti-cancer effects. Analogues resulting from simple functional group substitution at 3' position of VD3 were evaluated in a variety of biological assays to determine their ability to selectively inhibit Hh signaling. Moderately active Hh inhibitors that have insignificant binding affinity for VDR were identified; however, these compounds also activate the traditional VDR pathway, presumably due to metabolites produced in the cultured cells. Thus, further structural modifications to the VD3 scaffold are required to yield potent, selective Hh inhibitors.
Product Description
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
25-Hydroxyvitamin D3 solution, 5 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
25-Hydroxyvitamin D3 solution, 100 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
25-Hydroxyvitamin D3 solution, 50 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
25-Hydroxyvitamin D3 monohydrate, ≥99.0% (HPLC)
Sigma-Aldrich
25-Hydroxycholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 50 μg/mL in ethanol, 95% (CP)
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 100 μg/mL in ethanol, 95% (CP)