Merck
  • Impact of chromatin context on Cas9-induced DNA double-strand break repair pathway balance.
Impact of chromatin context on Cas9-induced DNA double-strand break repair pathway balance.
Molecular cell (2021-04-14)
Ruben Schep, Eva K Brinkman, Christ Leemans, Xabier Vergara, Robin H van der Weide, Ben Morris, Tom van Schaik, Stefano G Manzo, Daniel Peric-Hupkes, Jeroen van den Berg, Roderick L Beijersbergen, René H Medema, Bas van Steensel
ABSTRACT
DNA double-strand break (DSB) repair is mediated by multiple pathways. It is thought that the local chromatin context affects the pathway choice, but the underlying principles are poorly understood. Using a multiplexed reporter assay in combination with Cas9 cutting, we systematically measure the relative activities of three DSB repair pathways as a function of chromatin context in >1,000 genomic locations. This reveals that non-homologous end-joining (NHEJ) is broadly biased toward euchromatin, while the contribution of microhomology-mediated end-joining (MMEJ) is higher in specific heterochromatin contexts. In H3K27me3-marked heterochromatin, inhibition of the H3K27 methyltransferase EZH2 reverts the balance toward NHEJ. Single-stranded template repair (SSTR), often used for precise CRISPR editing, competes with MMEJ and is moderately linked to chromatin context. These results provide insight into the impact of chromatin on DSB repair pathway balance and guidance for the design of Cas9-mediated genome editing experiments.
Sigma-Aldrich
Monoclonal Anti-POLQ antibody produced in mouse, clone 1C11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Spermidine, BioReagent, Molecular Biology, suitable for cell culture, ≥98%
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Sigma-Aldrich
BIX 01294 trihydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Millipore
Digitonin, High Purity, CAS 11024-24-1 is a non-ionic detergent commonly used to solubilize membrane-bound proteins.