Merck
Bicephalic amphiphile architecture affects antibacterial activity.
European journal of medicinal chemistry (2011-07-29)
Jade E LaDow, David C Warnock, Kristina M Hamill, Kaitlin L Simmons, Robert W Davis, Christian R Schwantes, Devon C Flaherty, Jon A L Willcox, Kelsey Wilson-Henjum, Kevin L Caran, Kevin P C Minbiole, Kyle Seifert
ABSTRACT
A series of cationic amphiphiles, each with an aromatic core, was prepared and investigated for antimicrobial properties. The synthesized amphiphiles in this study are bicephalic (double-headed) in that they each possess two trimethylammonium head groups and a single linear alkoxy tail. Minimum inhibitory and minimum bactericidal concentrations of these amphiphiles were in the low micromolar range. Antimicrobial activities are highly sensitive to the chain length of the hydrophobic region, and modestly reliant on the relative positioning of the head groups on the aromatic core. These trends were more pronounced in time kill assays, wherein longer chain compounds required significantly shorter times to completely kill bacteria. Microscopy suggested that the mode of cell death was lysis. Strong inhibition was observed with both biscationic compounds and monocationic comparisons against Gram-positive bacteria; only biscationic amphiphiles maintained good activity versus the Gram-negative bacteria tested. These observations provide direction for future antimicrobial structural investigations.
Product Description
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Dodecyltrimethylammonium bromide, ≥98%
Supelco
Dodecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥98.5% (AT)
Sigma-Aldrich
Dodecyltrimethylammonium bromide, BioXtra, ~99%