Merck
  • Sodium dl-α-tocopheryl-6-O-phosphate inhibits PGE₂ production in keratinocytes induced by UVB, IL-1β and peroxidants.
Sodium dl-α-tocopheryl-6-O-phosphate inhibits PGE₂ production in keratinocytes induced by UVB, IL-1β and peroxidants.
Bioorganic & medicinal chemistry (2011-09-29)
Eiko Kato, Yuichi Sasaki, Noriko Takahashi
ABSTRACT
The water-soluble vitamin E derivative, sodium dl-α-tocopheryl-6-O-phosphate (1), exhibits protective effects against skin damage. As reported herein, we investigated the actions of 1 on the formation of the inflammatory mediator, prostaglandin E(2) (PGE(2)), as compared to dl-α-tocopheryl acetate (2) and dipotassium glycyrrhizin acid (3). In a three-dimensional (3D) human skin model 1 was converted to α-tocopherol (Toc) to a greater extent than 2. Post-treatment using 2% 1 following ultraviolet B (UVB) irradiation for 2h significantly reduced photodamage as indicated by UVB-damaged cell formation and PGE(2) synthesis. In normal human epidermal keratinocytes stimulated with UVB irradiation, or exposed to interleukin-1 beta, tert-butylhydroperoxide or hydrogen peroxide, pre-treatment with 1 (0-2 μM) inhibited PGE(2) production in dose-dependent manner to a greater extent than 2 and 3. Increases in stimulator-induced cyclooxygenase 2 mRNA expression and p38 MAPK phosphorylation were suppressed by pre-treatment with 1. The vitamin C derivative, magnesium L-ascorbyl-2-phosphate, significantly and synergistically, enhanced the inhibitory effects of 1 on PGE(2) production. These results suggest that 1 is a highly potent protective when compared among the examined commercial human skin care products, and that it might be useful for therapeutic and preventive medicine.
Sigma-Aldrich
(±)-α-Tocopherol phosphate disodium salt, water-soluble α-tocopherol analog
Sigma-Aldrich
DL-α-Tocopherol acetate, ≥96% (HPLC)
Sigma-Aldrich
DL-α-Tocopherol acetate, tested according to Ph. Eur.
Supelco
DL-alpha-Tocopherol acetate, analytical standard
Sigma-Aldrich
(+)-α-Tocopherol acetate, BioReagent, suitable for insect cell culture, ~1360 IU/g
Sigma-Aldrich
(+)-α-Tocopherol acetate, oil or semi-solid, ~1360 IU/g