Merck
  • GPR50-Ctail cleavage and nuclear translocation: a new signal transduction mode for G protein-coupled receptors.
GPR50-Ctail cleavage and nuclear translocation: a new signal transduction mode for G protein-coupled receptors.
Cellular and molecular life sciences : CMLS (2020-01-05)
Raise Ahmad, Olivier Lahuna, Anissa Sidibe, Avais Daulat, Qiang Zhang, Marine Luka, Jean-Luc Guillaume, Sarah Gallet, François Guillonneau, Juliette Hamroune, Sophie Polo, Vincent Prévot, Philippe Delagrange, Julie Dam, Ralf Jockers
ABSTRACT
Transmission of extracellular signals by G protein-coupled receptors typically relies on a cascade of intracellular events initiated by the activation of heterotrimeric G proteins or β-arrestins followed by effector activation/inhibition. Here, we report an alternative signal transduction mode used by the orphan GPR50 that relies on the nuclear translocation of its carboxyl-terminal domain (CTD). Activation of the calcium-dependent calpain protease cleaves off the CTD from the transmembrane-bound GPR50 core domain between Phe-408 and Ser-409 as determined by MALDI-TOF-mass spectrometry. The cytosolic CTD then translocates into the nucleus assisted by its 'DPD' motif, where it interacts with the general transcription factor TFII-I to regulate c-fos gene transcription. RNA-Seq analysis indicates a broad role of the CTD in modulating gene transcription with ~ 8000 differentially expressed genes. Our study describes a non-canonical, direct signaling mode of GPCRs to the nucleus with similarities to other receptor families such as the NOTCH receptor.
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Calpain Inhibitor II, powder
Sigma-Aldrich
Calpain Inhibitor I, ≥97% (TLC), powder
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human GTF2I
Sigma-Aldrich
Triton X-100, laboratory grade