Merck
  • Ursodeoxycholic acid amides as novel glucocorticoid receptor modulators.
Ursodeoxycholic acid amides as novel glucocorticoid receptor modulators.
Journal of medicinal chemistry (2010-12-17)
Ruchika Sharma, David Prichard, Ferenc Majer, Anne-Marie Byrne, Dermot Kelleher, Aideen Long, John F Gilmer
ABSTRACT
Ursodeoxycholic acid (UDCA) is used for the treatment of hepatic inflammatory diseases. Recent studies have shown that UDCA's biological effects are partly glucocorticoid receptor (GR) mediated. UDCA derivatives were synthesized and screened for ability to induce GR translocation in a high content analysis assay using the esophageal cancer SKGT-4 cell line. UDCA derivatives induced GR translocation in a time dependent manner with equal efficacy to that of dexamethasone (Dex) and with greatly increased potency relative to UDCA. The cyclopropylamide 1a suppressed TNF-α induced NF-κB activity and it induced GRE transactivation. 1a was unable to displace Dex from the GR ligand binding domain (LBD) in a competition experiment but was capable of coactivator recruitment in a time-resolved fluorescence energy transfer assay (TR-FRET). This represents a novel mechanism of action for a GR modulator. These derivatives could result in a new class of GR modulators.
Sigma-Aldrich
Ursodeoxycholic acid, ≥99%