Merck
  • Reduced histone deacetylase 7 activity restores function to misfolded CFTR in cystic fibrosis.
Reduced histone deacetylase 7 activity restores function to misfolded CFTR in cystic fibrosis.
Nature chemical biology (2009-12-08)
Darren M Hutt, David Herman, Ana P C Rodrigues, Sabrina Noel, Joseph M Pilewski, Jeanne Matteson, Ben Hoch, Wendy Kellner, Jeffery W Kelly, Andre Schmidt, Philip J Thomas, Yoshihiro Matsumura, William R Skach, Martina Gentzsch, John R Riordan, Eric J Sorscher, Tsukasa Okiyoneda, John R Yates, Gergely L Lukacs, Raymond A Frizzell, Gerard Manning, Joel M Gottesfeld, William E Balch
ABSTRACT
Chemical modulation of histone deacetylase (HDAC) activity by HDAC inhibitors (HDACi) is an increasingly important approach for modifying the etiology of human disease. Loss-of-function diseases arise as a consequence of protein misfolding and degradation, which lead to system failures. The DeltaF508 mutation in cystic fibrosis transmembrane conductance regulator (CFTR) results in the absence of the cell surface chloride channel and a loss of airway hydration, leading to the premature lung failure and reduced lifespan responsible for cystic fibrosis. We now show that the HDACi suberoylanilide hydroxamic acid (SAHA) restores surface channel activity in human primary airway epithelia to levels that are 28% of those of wild-type CFTR. Biological silencing of all known class I and II HDACs reveals that HDAC7 plays a central role in restoration of DeltaF508 function. We suggest that the tunable capacity of HDACs can be manipulated by chemical biology to counter the onset of cystic fibrosis and other human misfolding disorders.
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)