Merck
  • Quassidines A-D, bis-beta-carboline alkaloids from the stems of Picrasma quassioides.
Quassidines A-D, bis-beta-carboline alkaloids from the stems of Picrasma quassioides.
Journal of natural products (2010-01-26)
Wei-Hua Jiao, Hao Gao, Chen-Yang Li, Feng Zhao, Ren-Wang Jiang, Ying Wang, Guang-Xiong Zhou, Xin-Sheng Yao
ABSTRACT
Four new bis-beta-carboline alkaloids, quassidines A-D (1-4), together with a known alkaloid, picrasidine C (5), were isolated from the stems of Picrasma quassioides. Quassidine A (1) is the first reported bis-beta-carboline alkaloid possessing a novel cyclobutane moiety. The structures of the new compounds were determined on the basis of their 1D and 2D NMR and X-ray diffraction data. A possible biogenetic pathway for these alkaloids was proposed, and all compounds were evaluated for anti-inflammatory activity. Only quassidine A (1) showed weak activity.
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone-Water Soluble, BioReagent, suitable for cell culture