Merck
  • An efficient and economical MTT assay for determining the antioxidant activity of plant natural product extracts and pure compounds.
An efficient and economical MTT assay for determining the antioxidant activity of plant natural product extracts and pure compounds.
Journal of natural products (2010-06-23)
Yunbao Liu, Muraleedharan G Nair
ABSTRACT
Antioxidants scavenge free radicals, singlet oxygen, and electrons in cellular redox reactions. The yellow MTT [3-(4,5-dimethylthiazole-2-yl)-2,5-diphenyltetrazolium bromide] is reduced to a purple formazan by mitochondrial enzymes. NADPH is the basis of established in vitro cell viability assays. An antioxidant assay has been developed utilizing the redox reaction between MTT and selected natural product extracts and purified compounds. This simple, fast, and inexpensive MTT antioxidant assay is comparable with the lipid peroxidation inhibitory assay and can be mechanized to achieve high throughput.
Product Description
Supelco
Nonadecanoic acid, analytical standard
Supelco
Nonanoic acid, analytical standard
Supelco
Decanoic acid, analytical standard
Supelco
Behenic acid, analytical standard
Sigma-Aldrich
Myristic acid, ≥98.0% (GC)
Sigma-Aldrich
Arachidic acid, synthetic, ≥99.0% (GC)
Supelco
Octanoic acid, analytical standard
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Supelco
Erucic acid, analytical standard
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
(±)-Camphor, purum, synthetic, ≥95.0% (GC)
Sigma-Aldrich
Erucic acid, technical, ~90% (GC)
Sigma-Aldrich
Pyrogallol, ACS reagent
Sigma-Aldrich
Decanoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Nonanoic acid, ≥96%, FG
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Stearic acid 50, tested according to Ph. Eur.
Sigma-Aldrich
Camphor, 96%
Sigma-Aldrich
Myristic acid, ≥95%, FCC, FG
Sigma-Aldrich
Myristic acid, natural, ≥98.5%, FG
Sigma-Aldrich
(±)-Camphor, meets analytical specification of Ph. Eur., BP, ≥95% (GC), racemic
Sigma-Aldrich
(±)-Camphor, ≥95.5%
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Decanoic acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Octanoic acid, ≥98%, FG
Sigma-Aldrich
Behenic acid, 99%
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG