Merck
  • Chemical-based primary human hepatocyte monolayer culture for the study of drug metabolism and hepatotoxicity: Comparison with the spheroid model.
Chemical-based primary human hepatocyte monolayer culture for the study of drug metabolism and hepatotoxicity: Comparison with the spheroid model.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2021-02-11)
Yixiang Zhong, Jun Sang Yu, Xiaoqiong Wang, Bert Binas, Hye Hyun Yoo
ABSTRACT
Traditionally cultured monolayers of primary human hepatocytes (PHHs) deteriorate within days and thereby become unsuitable for drug-related studies. PHH spheroids (3D PHHs) maintain liver functions for weeks, but are considerably more demanding. Recently, a chemical-based approach (5C PHHs) succeeded in long-term culture of hepatocyte monolayers, but it remains unclear whether the drug-related functions are preserved. To clarify this, we compared the 5C and 3D PHHs in terms of gene expression analysis, proteomic analysis, functionality (basal and induced activities of representative CYP450 enzymes and urea and albumin secretions), survival in culture, and sensitivity to representative drugs. In all comparisons, which spanned culture durations of up to 4 weeks, the 5C PHHs performed at least as well as the 3D PHHs. Hence, the novel 5C PHH monolayer format combines the convenience of the traditional monolayer format with the functionality and maintainability of the spheroid format. Our results suggest that 5C PHH monolayers can be used more conveniently and efficiently for high-throughput drug screening, preclinical drug safety evaluations, and mechanistic studies.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Schiff′s reagent for aldehydes, suitable for microscopy
Sigma-Aldrich
Periodic acid, ACS reagent, 99%
Sigma-Aldrich
Diclofenac sodium salt
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid