- Comparative analysis of lentiviral gene transfer approaches designed to promote fetal hemoglobin production for the treatment of β-hemoglobinopathies.
Comparative analysis of lentiviral gene transfer approaches designed to promote fetal hemoglobin production for the treatment of β-hemoglobinopathies.
β-Hemoglobinopathies are among the most common single-gene disorders and are caused by different mutations in the β-globin gene. Recent curative therapeutic approaches for these disorders utilize lentiviral vectors (LVs) to introduce a functional copy of the β-globin gene into the patient's hematopoietic stem cells. Alternatively, fetal hemoglobin (HbF) can reduce or even prevent the symptoms of disease when expressed in adults. Thus, induction of HbF by means of LVs and other molecular approaches has become an alternative treatment of β-hemoglobinopathies. Here, we performed a head-to-head comparative analysis of HbF-inducing LVs encoding for: 1) IGF2BP1, 2) miRNA-embedded shRNA (shmiR) sequences specific for the γ-globin repressor protein BCL11A, and 3) γ-globin gene. Furthermore, two novel baboon envelope proteins (BaEV)-LVs were compared to the commonly used vesicular-stomatitis-virus glycoprotein (VSV-G)-LVs. Therapeutic levels of HbF were achieved for all VSV-G-LV approaches, from a therapeutic level of 20% using γ-globin LVs to 50% for both IGF2BP1 and BCL11A-shmiR LVs. Contrarily, BaEV-LVs conferred lower HbF expression with a peak level of 13%, however, this could still ameliorate symptoms of disease. From this thorough comparative analysis of independent HbF-inducing LV strategies, we conclude that HbF-inducing VSV-G-LVs represent a promising alternative to β-globin gene addition for patients with β-hemoglobinopathies.