Merck
  • Ubiquitin-independent proteasomal degradation of Spindlin-1 by the E3 ligase HACE1 contributes to cell-cell adhesion.
Ubiquitin-independent proteasomal degradation of Spindlin-1 by the E3 ligase HACE1 contributes to cell-cell adhesion.
FEBS letters (2021-01-10)
Vivek Reddy Palicharla, Devanshi Gupta, Debjani Bhattacharya, Subbareddy Maddika
ABSTRACT
HECT-E3 ligases play an essential role in catalyzing the transfer of ubiquitin to protein substrates. The noncatalytic roles of HECT-E3 ligases in cells are unknown. Here, we report that a HECT-E3 ligase, HACE1, functions as an adaptor independent of its E3 ligase activity. We identified Spindlin-1, a histone reader, as a new HACE1-associated protein. Interestingly, we found that HACE1 promotes Spindlin-1 degradation via the proteasome in an ubiquitination-independent manner. Functionally, we demonstrated that the loss of HACE1 results in weak cell-cell adhesion due to Spindlin-1-mediated accumulation of GDNF, a negative regulator of cell adhesion. Together, our data suggest that HACE1 acts as a molecular adaptor and plays an important noncatalytic role in presenting selected substrates directly to the proteasome for degradation.
Sigma-Aldrich
Monoclonal Anti-Maltose Binding Protein antibody produced in mouse, clone MBP-17, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
MISSION® esiRNA, targeting human SPIN1
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Maltose solution, BioReagent, for molecular biology, ~20% in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human HACE1
Sigma-Aldrich
Anti-Ubiquitin Antibody, clone P4D1-A11, clone P4D1-A11, Upstate®, from mouse