Merck
  • Enzyme functionalized microgels enable precise regulation of dissolved oxygen and anaerobe culture.
Enzyme functionalized microgels enable precise regulation of dissolved oxygen and anaerobe culture.
Materials today. Bio (2021-02-09)
A S Jeevarathinam, F Guo, T Williams, J A Smolen, J A Hyde, M J McShane, P de Figueiredo, D L Alge
ABSTRACT
Anaerobes are a major constituent of the gut microbiome and profoundly influence the overall health of humans. However, the lack of a simple, cost-effective, and scalable system that mimics the anaerobic conditions of the human gut is hindering research on the gut microbiome and the development of therapeutics. Here, we address this gap by using glucose oxidase and catalase containing gelatin microparticles (GOx-CAT-GMPs) to precisely regulate dissolved oxygen concentration [O2] via GOx-mediated consumption of oxygen. Fluorescence images generated using conjugated polymer afterglow nanoparticles showed that [O2] can be tuned from 257.9 ​± ​6.2 to 0.0 ​± ​4.0 ​μM using GOx-CAT-GMPs. Moreover, when the obligate anaerobe Bacteroides thetaiotaomicron was inoculated in media containing GOx-CAT-GMPs, bacterial growth under ambient oxygen was comparable to control conditions in an anaerobic chamber (5.4 ​× ​105 and 8.8 ​× ​105 colony forming units mL-1, respectively). Finally, incorporating GOx-CAT-GMPs into a bioreactor that permitted continuous radial diffusion of oxygen and glucose generated a gut-mimetic [O2] gradient of 132.4 ​± ​2.6 ​μM in the outer ring of the reactor to 7.9 ​± ​1.7 ​μM at the core. Collectively, these results indicate that GOx-CAT-GMPs are highly effective oxygen-regulating materials. These materials can potentially be leveraged to advance gut microbiome research and fecal microbiota transplantation, particularly in low-resource settings.
Sigma-Aldrich
Poly[2-methoxy-5-(2-ethylhexyloxy)-1,4-phenylenevinylene], average Mn 40,000-70,000
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Hematin porcine
Millipore
Brain Heart Infusion Broth, NutriSelect® Plus, suitable for microbiology
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~14,600
Sigma-Aldrich
Vitamin K1, BioXtra, ≥99.0% (sum of isomers, HPLC), mixture of isomers
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A