Merck
Synthesis and anticancer evaluation of alpha-lipoic acid derivatives.
Bioorganic & medicinal chemistry letters (2010-04-21)
Shi-Jie Zhang, Qiu-Fu Ge, Dian-Wu Guo, Wei-Xiao Hu, Hua-Zhang Liu
ABSTRACT
alpha-Lipoic acid derivatives were synthesized and evaluated for their in vitro anticancer activities against NCI-460, HO-8910, KB, BEL-7402, and PC-3 cell lines. The results, for most compounds exhibited dose-dependent inhibitory property and several compounds had good inhibitions at the dose of 100 microg/mL. Compound 17 m was further selected for in vivo evaluation against S180 xenograft in ICR mice, which had 24.7% tumor-weight inhibition through intragastric administration of 200mg/kg of body weight. Moreover, the LD(50) in mice for 17 m through ig exceeded 1000 mg/kg of body weight.
Sigma-Aldrich
(±)-α-Lipoic acid, ≥98.0%
Sigma-Aldrich
(±)-α-Lipoic acid, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
(±)-α-Lipoic acid, synthetic, ≥99% (titration), powder