Merck
  • Erb-b2 Receptor Tyrosine Kinase 2 (ERBB2) Promotes ATG12-Dependent Autophagy Contributing to Treatment Resistance of Breast Cancer Cells.
Erb-b2 Receptor Tyrosine Kinase 2 (ERBB2) Promotes ATG12-Dependent Autophagy Contributing to Treatment Resistance of Breast Cancer Cells.
Cancers (2021-04-04)
Yongqiang Chen, Ruobing Wang, Shujun Huang, Elizabeth S Henson, Jayce Bi, Spencer B Gibson
ABSTRACT
The epidermal growth factor receptor (EGFR) family member erb-b2 receptor tyrosine kinase 2 (ERBB2) is overexpressed in many types of cancers leading to (radio- and chemotherapy) treatment resistance, whereas the underlying mechanisms are still unclear. Autophagy is known to contribute to cancer treatment resistance. In this study, we demonstrate that ERBB2 increases the expression of different autophagy genes including ATG12 (autophagy-related 12) and promotes ATG12-dependent autophagy. We clarify that lapatinib, a dual inhibitor for EGFR and ERBB2, promoted autophagy in cells expressing only EGFR but inhibited autophagy in cells expressing only ERBB2. Furthermore, breast cancer database analysis of 35 genes in the canonical autophagy pathway shows that the upregulation of ATG12 and MAP1LC3B is associated with a low relapse-free survival probability of patients with ERBB2-positive breast tumors following treatments. Downregulation of ERBB2 or ATG12 increased cell death induced by chemotherapy drugs in ERBB2-positive breast cancer cells, whereas upregulation of ERBB2 or ATG12 decreased the cell death in ERBB2-negative breast cancer cells. Finally, ERBB2 antibody treatment led to reduced expression of ATG12 and autophagy inhibition increasing drug or starvation-induced cell death in ERBB2-positive breast cancer cells. Taken together, this study provides a novel approach for the treatment of ERBB2-positive breast cancer by targeting ATG12-dependent autophagy.
Sigma-Aldrich
MISSION® esiRNA, targeting human ATG12
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
IGEPAL® CA-630
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Anti-Actin antibody, Mouse monoclonal, clone AC-40, purified from hybridoma cell culture