Merck
  • Polyamines inhibit carbonic anhydrases by anchoring to the zinc-coordinated water molecule.
Polyamines inhibit carbonic anhydrases by anchoring to the zinc-coordinated water molecule.
Journal of medicinal chemistry (2010-07-02)
Fabrizio Carta, Claudia Temperini, Alessio Innocenti, Andrea Scozzafava, Kai Kaila, Claudiu T Supuran
ABSTRACT
Carbonic anhydrases (CAs, EC 4.2.1.1) are inhibited by sulfonamides, phenols, and coumarins. Polyamines such as spermine, spermidine, and many synthetic congeners are described to constitute a novel class of CA inhibitors (CAIs), interacting with the different CA isozymes with efficiency from the low nanomolar to millimolar range. The main structure-activity relationship for these CAIs have been delineated: the length of the molecule, number of amine moieties, and their functionalization are the main parameters controlling activity. The X-ray crystal structure of the CA II-spermine adduct allowed understanding of the inhibition mechanism. Spermine anchors to the nonprotein zinc ligand through a network of hydrogen bonds. Its distal amine moiety makes hydrogen bonds with residues Thr200 and Pro201, which further stabilize the adduct. Spermine binds differently compared to sulfonamides, phenols, or coumarins, rendering possible to develop CAIs with a diverse inhibition mechanism, profile, and selectivity for various isoforms.
Sigma-Aldrich
Cadaverine, 95%
Sigma-Aldrich
Diethylenetriamine, ReagentPlus®, 99%
Sigma-Aldrich
Hexamethylenediamine, technical grade, 70%
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
N,N′-Bis(2-aminoethyl)-1,3-propanediamine, 97%
Sigma-Aldrich
Hexamethylenediamine, 98%
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Spermine, BioReagent, suitable for cell culture
Sigma-Aldrich
Spermine, ≥97%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
1,1,4,7,10,10-Hexamethyltriethylenetetramine, 97%
Sigma-Aldrich
Triethylenetetramine, technical grade, 60%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Supelco
Phenol, PESTANAL®, analytical standard
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
Triethylenetetramine, ≥97.0% (T)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Spermidine, BioReagent, for molecular biology, suitable for cell culture, ≥98%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
Spermidine, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ≥99%