Merck
  • Kinesin spindle protein (KSP) inhibitors with 2,3-fused indole scaffolds.
Kinesin spindle protein (KSP) inhibitors with 2,3-fused indole scaffolds.
Journal of medicinal chemistry (2010-06-05)
Shinya Oishi, Toshiaki Watanabe, Jun-Ichi Sawada, Akira Asai, Hiroaki Ohno, Nobutaka Fujii
ABSTRACT
Mitotic kinesin spindle protein (KSP) is involved in the assembly of the bipolar spindle during cell division. On the basis of a common 2,3-fused indole substructure within the complex frameworks of terpendole E and other KSP inhibitors, the carbazoles with a bulky alkyl group were identified as a novel KSP inhibitory scaffold. Additionally, among several naturally occurring cell growth inhibitors with 2,3-fused indole structures, beta-carboline alkaloids, harman and harmine, showed moderate inhibition of KSP.
Product Description
Supelco
Carbazole, VETRANAL®, analytical standard
Sigma-Aldrich
Harmine, 98%
Sigma-Aldrich
2-Hydroxycarbazole, 97%
Sigma-Aldrich
Carbazole, ≥95% (GC)