Merck
  • N-glycosylation shields Phytophthora sojae apoplastic effector PsXEG1 from a specific host aspartic protease.
N-glycosylation shields Phytophthora sojae apoplastic effector PsXEG1 from a specific host aspartic protease.
Proceedings of the National Academy of Sciences of the United States of America (2020-10-22)
Yeqiang Xia, Zhenchuan Ma, Min Qiu, Baodian Guo, Qi Zhang, Haibin Jiang, Baiyu Zhang, Yachun Lin, Mingrun Xuan, Liang Sun, Haidong Shu, Junhua Xiao, Wenwu Ye, Yan Wang, Yiming Wang, Suomeng Dong, Brett M Tyler, Yuanchao Wang
ABSTRACT
Hosts and pathogens are engaged in a continuous evolutionary struggle for physiological dominance. A major site of this struggle is the apoplast. In Phytophthora sojae-soybean interactions, PsXEG1, a pathogen-secreted apoplastic endoglucanase, is a key focal point of this struggle, and the subject of two layers of host defense and pathogen counterdefense. Here, we show that N-glycosylation of PsXEG1 represents an additional layer of this coevolutionary struggle, protecting PsXEG1 against a host apoplastic aspartic protease, GmAP5, that specifically targets PsXEG1. This posttranslational modification also attenuated binding by the previously described host inhibitor, GmGIP1. N-glycosylation of PsXEG1 at N174 and N190 inhibited binding and degradation by GmAP5 and was essential for PsXEG1's full virulence contribution, except in GmAP5-silenced soybeans. Silencing of GmAP5 reduced soybean resistance against WT P. sojae but not against PsXEG1 deletion strains of P. sojae. The crucial role of N-glycosylation within the three layers of defense and counterdefense centered on PsXEG1 highlight the critical importance of this conserved apoplastic effector and its posttranslational modification in Phytophthora-host coevolutionary conflict.
Sigma-Aldrich
Pepstatin A, microbial, ≥75% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for plant cell and tissue extracts, DMSO solution
Sigma-Aldrich
1,10-Phenanthroline monohydrate, reagent grade
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
Bestatin hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
E-64, protease inhibitor