Merck
  • Enhanced UHPLC-MS/MS screening of selective androgen receptor modulators following urine hydrolysis.
Enhanced UHPLC-MS/MS screening of selective androgen receptor modulators following urine hydrolysis.
MethodsX (2020-06-18)
Anna Gadaj, Emiliano Ventura, Jim Healy, Francesco Botrè, Saskia S Sterk, Tom Buckley, Mark H Mooney
ABSTRACT
Selective androgen receptor modulators (SARMs) represent non-steroidal agents commonly abused in human and animal (i.e. equine, canine) sports, with potential for further misuse as growth promoting agents in livestock-based farming. As a direct response to the real and possible implications of illicit application in both sport as well as food production systems, this study incorporated enzymatic hydrolysis (β-glucuronidase/arylsulfatase) into a previously established protocol while maintaining the minimal volume (200 µL) of urine sample required to detect SARMs encompassing various pharmacophores in urine from a range of species (i.e. equine, bovine, human, canine and rodent). The newly presented semi-quantitative UHPLC-MS/MS-based assay is shown to be fit-for-purpose, being rapid and offering high-throughput, with validation findings fulfilling criteria stipulated within relevant doping and food control legislation.•CCβ values determined at 1 ng mL-1 for majority of analytes.•Deconjugation step included in the method led to significantly increased relative abundance of ostarine in analysed incurred urine samples demonstrating the requirement for hydrolysis to detect a total form of emerging SARMs.•Assay amenable for use within routine testing to ensure fair play in animal and human sports and that animal-derived food is free from contamination with SARM residues.
Product Description
Supelco
6-[Bis(2,2,2-trifluoroethyl)amino]-4-(trifluoromethyl)-2(1H)-quinolinone, analytical standard