Merck
  • ROS1-fusion protein induces PD-L1 expression via MEK-ERK activation in non-small cell lung cancer.
ROS1-fusion protein induces PD-L1 expression via MEK-ERK activation in non-small cell lung cancer.
Oncoimmunology (2020-09-15)
Zheng Liu, Kejia Zhao, Shiyou Wei, Chengwu Liu, Jiankang Zhou, Qiheng Gou, Xia Wu, Zhenyu Yang, Yanbo Yang, Yong Peng, Qing Cheng, Lunxu Liu
ABSTRACT
Despite some of the oncogenic driver mutations that have been associated with increased expression of programmed death-ligand 1 (PD-L1), the correlation between PD-L1 expression and ROS1 fusion in NSCLC cells, especially for those with Crizotinib resistance has not been fully addressed. The expression of PD-L1 in 30 primary NSCLC tumors with/without ROS1-fusion protein was evaluated by immunohistochemical (IHC) analysis. To assess the correlation between ROS1 fusion and PD-L1 expression, we down-regulated ROS1 with RNA interference or specific inhibitor (Crizotinib) in ROS1-fusion positive NSCLC cell line HCC78; or up-regulate ROS1-fusion gene in an immortalized human bronchial epithelial cell line (HBE). Mouse xenograft models were also used to determine the effect of ROS1 expression on PD-L1 expression in vivo. Crizotinib-resistant cell line was generated for measuring the association between Crizotinib resistance and PD-L1 expression. ROS1-rearrangement in primary NSCLC tumor was significantly associated with up-regulated PD-L1 expression. PD-L1 expression was significantly up-regulated in bronchial epithelial cells after forced expression of ROS1 fusion and was eliminated when HCC78 xenograft mouse models were treated with Crizotinib. We found PD-L1 expression was modulated by MEK-ERK pathway signaling in both parental and Crizotinib-resistant NSCLC cells with ROS1 fusion. The correlation between ROS1-fusion and PD-L1 overexpression suggested that PD-L1/PD-1 blockade could be the second-line treatment option for the Crizotinib-resistant NSCLC with ROS1 rearrangement.
Sigma-Aldrich
Crizotinib, ≥98% (HPLC)