Merck
The neural basis for a persistent internal state in Drosophila females.
eLife (2020-11-24)
David Deutsch, Diego Pacheco, Lucas Encarnacion-Rivera, Talmo Pereira, Ramie Fathy, Jan Clemens, Cyrille Girardin, Adam Calhoun, Elise Ireland, Austin Burke, Sven Dorkenwald, Claire McKellar, Thomas Macrina, Ran Lu, Kisuk Lee, Nico Kemnitz, Dodham Ih, Manuel Castro, Akhilesh Halageri, Chris Jordan, William Silversmith, Jingpeng Wu, H Sebastian Seung, Mala Murthy
ABSTRACT
Sustained changes in mood or action require persistent changes in neural activity, but it has been difficult to identify the neural circuit mechanisms that underlie persistent activity and contribute to long-lasting changes in behavior. Here, we show that a subset of Doublesex+ pC1 neurons in the Drosophila female brain, called pC1d/e, can drive minutes-long changes in female behavior in the presence of males. Using automated reconstruction of a volume electron microscopic (EM) image of the female brain, we map all inputs and outputs to both pC1d and pC1e. This reveals strong recurrent connectivity between, in particular, pC1d/e neurons and a specific subset of Fruitless+ neurons called aIPg. We additionally find that pC1d/e activation drives long-lasting persistent neural activity in brain areas and cells overlapping with the pC1d/e neural network, including both Doublesex+ and Fruitless+ neurons. Our work thus links minutes-long persistent changes in behavior with persistent neural activity and recurrent circuit architecture in the female brain.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
all trans-Retinal, powder, ≥98%
Sigma-Aldrich
Schneider′s Insect Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for insect cell culture
Millipore
Universal Food pathogen ID Membrane, light pink colored, sterile, diameter 70 mm, suitable for microbiology