Merck
  • Correction to: Brain-derived exosomes from dementia with Lewy bodies propagate α-synuclein pathology.
Correction to: Brain-derived exosomes from dementia with Lewy bodies propagate α-synuclein pathology.
Acta neuropathologica communications (2020-08-06)
Jennifer Ngolab, Ivy Trinh, Edward Rockenstein, Michael Mante, Jazmin Florio, Margarita Trejo, Deborah Masliah, Anthony Adame, Eliezer Masliah, Robert A Rissman
ABSTRACT
An amendment to this paper has been published and can be accessed via the original article.