Merck
  • Wake-sleep cycles are severely disrupted by diseases affecting cytoplasmic homeostasis.
Wake-sleep cycles are severely disrupted by diseases affecting cytoplasmic homeostasis.
Proceedings of the National Academy of Sciences of the United States of America (2020-10-28)
Stephen Beesley, Dae Wook Kim, Matthew D'Alessandro, Yuanhu Jin, Kwangjun Lee, Hyunjeong Joo, Yang Young, Robert J Tomko, John Faulkner, Joshua Gamsby, Jae Kyoung Kim, Choogon Lee
ABSTRACT
The circadian clock is based on a transcriptional feedback loop with an essential time delay before feedback inhibition. Previous work has shown that PERIOD (PER) proteins generate circadian time cues through rhythmic nuclear accumulation of the inhibitor complex and subsequent interaction with the activator complex in the feedback loop. Although this temporal manifestation of the feedback inhibition is the direct consequence of PER's cytoplasmic trafficking before nuclear entry, how this spatial regulation of the pacemaker affects circadian timing has been largely unexplored. Here we show that circadian rhythms, including wake-sleep cycles, are lengthened and severely unstable if the cytoplasmic trafficking of PER is disrupted by any disease condition that leads to increased congestion in the cytoplasm. Furthermore, we found that the time delay and robustness in the circadian clock are seamlessly generated by delayed and collective phosphorylation of PER molecules, followed by synchronous nuclear entry. These results provide clear mechanistic insight into why circadian and sleep disorders arise in such clinical conditions as metabolic and neurodegenerative diseases and aging, in which the cytoplasm is congested.
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Oil Red O, certified by the Biological Stain Commission
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
Earle′s Balanced Salts, With sodium bicarbonate, without calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)
Sigma-Aldrich
MG-132, A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).