Merck
  • Tight Junction Protein Claudin-12 Is Involved in Cell Migration during Metastasis.
Tight Junction Protein Claudin-12 Is Involved in Cell Migration during Metastasis.
Biomolecules (2021-05-01)
Desislava Kolchakova, Dzhemal Moten, Tsvetelina Batsalova, Balik Dzhambazov
ABSTRACT
Claudins are important components of the tight junctions determining barrier properties, cell polarity, and paracellular permeability. Although many functions of claudins in cancer cells have not been elucidated, recent studies have shown that claudins play an important role in cell migration and metastasis. Loss of epithelial/endothelial integrity, disruption of tight junctions, and increased paracellular leakage are often observed during metastasis. The aim of our study was to investigate the involvement of claudin-12 in the process of cell migration as well as to evaluate the possibility of using this protein as a specific target for the regulation of tumorigenesis. We have performed immunocytochemistry assays to detect the expression of claudin-12 in different epithelial/endothelial human cell lines, and selected three (A549, LS180, and HeLa) for further experiments. Using transwell chamber migration assays, we found that anti-claudin-12 antibodies inhibited both the migration and proliferation of claudin-12 expressing cells (A549 and LS180), inducing apoptosis, as well as the migration capacity of Jurkat cells through the monolayers formed from A549 or LS180 cells. In addition, co-cultures of Jurkat cells on monolayers from A549 or LS180 cells, in the presence of synthetic claudin-12 peptides representing the extracellular domains of the claudin-12 protein, also reduced the number of migrated Jurkat cells. Two of the tested peptides (p5 and p6) almost completely blocked the migration of Jurkat cells. All migrated Jurkat cells expressed LFA-1 and CD62L, but not CD44. Thus, claudin-12 is a suitable biomarker for tumor progression and metastasis and an attractive target for antitumor therapy. Anti-claudin-12 antibodies and competitive inhibitory peptides could be useful in the therapeutic approach applied to cancer metastasis in tissues expressing claudin-12.
Sigma-Aldrich
Monoclonal Anti-CLDN12 antibody produced in mouse, clone 2D8, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution