Merck
  • Synthesis and preliminary antibacterial evaluation of simplified thiomarinol analogs.
Synthesis and preliminary antibacterial evaluation of simplified thiomarinol analogs.
Bioorganic & medicinal chemistry (2008-02-01)
Olivier Marion, Xuri Gao, Sandra Marcus, Dennis G Hall
ABSTRACT
A series of simplified thiomarinol derivatives was prepared by way of catalytic enantioselective inverse electron demand hetero [4+2] cycloaddition/allylboration tandem reaction. As a preliminary evaluation, these analogs were tested for antimicrobial activity using a standard disk diffusion assay. Whereas amide analogs were less active, the simple ester analogs 3 and 4 were demonstrated to be more active than thiomarinol H.
Sigma-Aldrich
Penicillin G sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Penicillin G sodium salt, ~1650 U/mg
Supelco
Tetracycline hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
Sigma-Aldrich
Tetracycline hydrochloride, powder