Merck
  • Gelatin device for the delivery of growth factors involved in endochondral ossification.
Gelatin device for the delivery of growth factors involved in endochondral ossification.
PloS one (2017-04-06)
Lucas A J Ahrens, Daniel Vonwil, Jon Christensen, V Prasad Shastri
ABSTRACT
Controlled release drug delivery systems are well established as oral and implantable dosage forms. However, the controlled release paradigm can also be used to present complex soluble signals responsible for cellular organization during development. Endochondral ossification (EO), the developmental process of bone formation from a cartilage matrix is controlled by several soluble signals with distinct functions that vary in structure, molecular weight and stability. This makes delivering them from a single vehicle rather challenging. Herein, a gelatin-based delivery system suitable for the delivery of small molecules as well as recombinant human (rh) proteins (rhWNT3A, rhFGF2, rhVEGF, rhBMP4) is reported. The release behavior and biological activity of the released molecules was validated using analytical and biological assays, including cell reporter systems. The simplicity of fabrication of the gelatin device should foster its adaptation by the diverse scientific community interested in interrogating developmental processes, in vivo.
Sigma-Aldrich
Fluorometric Aldehyde Assay Kit, sufficient for 200 fluorometric tests
Sigma-Aldrich
Wnt Agonist, The Wnt Agonist, also referenced under CAS 853220-52-7, controls the biological activity of Wnt. This small molecule/inhibitor is primarily used for Cancer applications.