Merck
  • Effects of Dietary Carbohydrate Content on Circulating Metabolic Fuel Availability in the Postprandial State.
Effects of Dietary Carbohydrate Content on Circulating Metabolic Fuel Availability in the Postprandial State.
Journal of the Endocrine Society (2020-07-16)
Kim J Shimy, Henry A Feldman, Gloria L Klein, Lisa Bielak, Cara B Ebbeling, David S Ludwig
ABSTRACT
According to the carbohydrate-insulin model of obesity, an elevated insulin-to-glucagon ratio in response to a high-carbohydrate diet directs metabolic fuels toward storage, resulting in lower circulating energy. To determine differences in total circulating energy post-meal related to dietary carbohydrate. Ancillary study within the Framingham State Food Study. University community. 29 adults (aged 20 to 65 years) with overweight or obesity (body mass index ≥25 kg/m2). After achieving 10% to 14% weight loss on a run-in diet, participants were randomized to weight-loss-maintenance test diets varying in carbohydrate content (high-carbohydrate, 60% of total energy, n = 11; moderate-carbohydrate, 40%, n = 8; low-carbohydrate, 20%, n = 10) and controlled for protein (20%). During 24-hour metabolic ward admissions between 10 and 15 weeks on the test diets, metabolic fuels and hormones were measured. Energy availability (EA) based on energy content of blood glucose, beta-hydroxybutyrate, and free fatty acids, in the late postprandial period (180 to 300 minutes). Insulin at 30 minutes into the test meal (Meal Insulin-30) was measured as an effect modifier. Insulin-to-glucagon ratio was 7-fold higher in participants on the high- vs low-carbohydrate diet (2.5 and 0.36, respectively). Late postprandial EA was 0.58 kcal/L lower on the high- vs low-carbohydrate diet (P < 0.0001), primarily related to suppression of free fatty acids. Early postprandial EA (30 to 180 minutes) declined fastest in the high-carbohydrate group, and Meal Insulin-30 modified this diet effect. During weight-loss maintenance on a high-carbohydrate diet, late postprandial EA is reduced, consistent with the carbohydrate-insulin model.
Sigma-Aldrich
Ketone Body Assay, sufficient for 200 tests (UV)