Merck
  • Relation between lipophilicity of alkyl gallates and antifungal activity against yeasts and filamentous fungi.
Relation between lipophilicity of alkyl gallates and antifungal activity against yeasts and filamentous fungi.
Bioorganic & medicinal chemistry letters (2009-02-24)
P C Leal, A Mascarello, M Derita, F Zuljan, R J Nunes, S Zacchino, R A Yunes
ABSTRACT
The antifungal activity of a complete series of 15 n-alkyl gallates and six analogues acting against a representative panel of opportunistic pathogenic fungi was studied in order to analyze their role in: the importance of the fungi tested, the importance of the hydroxyls, the influence of the chain length and the hydrophobicity of the compounds. It was demonstrated that dermatophytes were the most susceptible species and that hydroxyls appear to be necessary but not sufficient for the activity. When the logP of each gallate was calculated and related to the different values of MIC against Microsporum gypseum it was observed that hexyl, heptyl, octyl and nonyl gallates exhibit a significant positive deviation from the curve corresponding to a polynomial equation obtained for the other gallates. This suggests that these compounds have a further mode of action besides their hydrophobicity, possibly the inhibition of some enzyme involved in ergosterol biosynthesis.
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Gallic acid monohydrate, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Pyrogallol, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Gallic acid monohydrate, ACS reagent, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Methyl 3,4,5-trihydroxybenzoate, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
3,4,5-Trimethoxybenzoic acid, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Lauryl gallate, antioxidant, ≥99.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Propyl gallate, antioxidant, ≥98.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Propyl gallate, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Propyl gallate, ≥98%, FCC
Pricing and availability is not currently available.