Merck
  • Neuroprotective and cholinergic properties of multifunctional glutamic acid derivatives for the treatment of Alzheimer's disease.
Neuroprotective and cholinergic properties of multifunctional glutamic acid derivatives for the treatment of Alzheimer's disease.
Journal of medicinal chemistry (2009-10-28)
Mariana P Arce, María Isabel Rodríguez-Franco, Gema C González-Muñoz, Concepción Pérez, Beatriz López, Mercedes Villarroya, Manuela G López, Antonio G García, Santiago Conde
ABSTRACT
Novel multifunctional compounds have been designed, synthesized, and evaluated as potential drugs for the treatment of Alzheimer's disease (AD). With an L-glutamic moiety as a suitable biocompatible linker, three pharmacophoric groups were joined: (1) an N-benzylpiperidine fragment selected to inhibit acetylcholinesterase by interacting with the catalytic active site (CAS), (2) an N-protecting group of the amino acid, capable of interacting with the acetylcholinesterase (AChE)-peripheral anionic site (PAS) and protecting neurons against oxidative stress, and (3) a lipophilic alkyl ester that would facilitate penetration into the central nervous system by crossing the blood-brain barrier. At submicromolar concentration, they inhibit AChE and butyrylcholinesterase (BuChE) of human origin, displace the binding of propidium iodide from the PAS of AChE, and could thus inhibit Abeta aggregation promoted by AChE. They also display neuroprotective properties against mitochondrial free radicals, show low toxicity, and could be able to penetrate into the CNS.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Progesterone, VETRANAL®, analytical standard
Supelco
Ofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Supelco
Testosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Piroxicam, meets USP testing specifications
Sigma-Aldrich
Progesterone-Water Soluble, powder, BioReagent, suitable for cell culture