Merck
  • Lipophilicity of basic drugs measured by hydrophilic interaction chromatography.
Lipophilicity of basic drugs measured by hydrophilic interaction chromatography.
Journal of medicinal chemistry (2009-04-29)
Bruno Bard, Pierre-Alain Carrupt, Sophie Martel
ABSTRACT
RPLC gains acceptance in pharmaceutical research for the rapid determination of lipophilicity but remains limited for the determination of partition coefficients of moderate to strong basic compounds under their neutral form because stationary phases are not compatible with high pH conditions. In this work, HILIC technique was used to accurately measure log P(oct) of the neutral form of basic drugs by measuring the difference between 2 isocratic log k values (Delta log k(0-95)) of their cationic form.
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
(1S,2S)-(+)-Pseudoephedrine, 98%
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
N,N-Dimethylbenzylamine, ≥99%
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Sigma-Aldrich
Morpholine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Butylamine, ≥99%
Supelco
Carazolol, analytical standard
Sigma-Aldrich
L-Tyrosine methyl ester, 98%
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Pindolol, ≥98% (TLC), powder
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Ethanolamine, ≥99%
Supelco
Ethanolamine, analytical standard
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Supelco
Morpholine, analytical standard
Sigma-Aldrich
Morpholine, ACS reagent, ≥99.0%
Sigma-Aldrich
Morpholine, ReagentPlus®, ≥99%
Sigma-Aldrich
Butylamine, 99.5%