Merck
  • Discovery and characterization of a marine bacterial SAM-dependent chlorinase.
Discovery and characterization of a marine bacterial SAM-dependent chlorinase.
Nature chemical biology (2007-12-07)
Alessandra S Eustáquio, Florence Pojer, Joseph P Noel, Bradley S Moore
ABSTRACT
Halogen atom incorporation into a scaffold of bioactive compounds often amplifies biological activity, as is the case for the anticancer agent salinosporamide A (1), a chlorinated natural product from the marine bacterium Salinispora tropica. Significant effort in understanding enzymatic chlorination shows that oxidative routes predominate to form reactive electrophilic or radical chlorine species. Here we report the genetic, biochemical and structural characterization of the chlorinase SalL, which halogenates S-adenosyl-L-methionine (2) with chloride to generate 5'-chloro-5'-deoxyadenosine (3) and L-methionine (4) in a rarely observed nucleophilic substitution strategy analogous to that of Streptomyces cattleya fluorinase. Further metabolic tailoring produces a halogenated polyketide synthase substrate specific for salinosporamide A biosynthesis. SalL also accepts bromide and iodide as substrates, but not fluoride. High-resolution crystal structures of SalL and active site mutants complexed with substrates and products support the S(N)2 nucleophilic substitution mechanism and further illuminate halide specificity in this newly discovered halogenase family.
Product Description
SAFC
L-Methionine
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)