Merck
  • [6]-gingerol induces Ca2+ mobilization in Madin-Darby canine kidney cells.
[6]-gingerol induces Ca2+ mobilization in Madin-Darby canine kidney cells.
Journal of natural products (2008-01-10)
Chung-Yi Chen, Ching-Hsein Chen, Chiu-Hu Kung, Shih-Hsing Kuo, Soong-Yu Kuo
ABSTRACT
[6]-gingerol, a major phenolic compound derived from ginger (Zingiber officinale), is a potential chemopreventive compound that can induce stress in cancer cells and cause apoptotic cell death. This study examines the early signaling effects of [6]-gingerol on renal cells. It was found that [6]-gingerol caused a slow and sustained rise of [Ca2+]i in a concentration-dependent manner. [6]-gingerol also induced a [Ca2+]i rise when extracellular Ca2+ was removed, but the magnitude was reduced by 80%. Depletion of intracellular Ca2+ stores with CCCP, a mitochondrial uncoupler, did not affect the action of [6]-gingerol. In a Ca2+-free medium, the [6]-gingerol-induced [Ca2+]i rise was partially abolished by depleting stored Ca2+ with thapsigargin (an endoplasmic reticulum Ca2+ pump inhibitor). The elevation of [6]-gingerol-caused [Ca2+]i in a Ca2+-containing medium was not affected by modulation of protein kinase C activity. The [6]-gingerol-induced Ca2+ influx was blocked by nicardipine. U73122, an inhibitor of phospholipase C, abolished ATP (but not [6]-gingerol)-induced [Ca2+]i rise. These findings suggest that [6]-gingerol induces a significant rise in [Ca2+]i in MDCK renal tubular cells by stimulating both extracellular Ca2+ influx and thapsigargin-sensitive intracellular Ca2+ release via as yet unidentified mechanisms.
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Supelco
[6]-Gingerol, analytical standard
Sigma-Aldrich
[6]-Gingerol, ≥98% (HPLC)