Merck
Hhip haploinsufficiency sensitizes mice to age-related emphysema.
Proceedings of the National Academy of Sciences of the United States of America (2016-07-23)
Taotao Lao, Zhiqiang Jiang, Jeong Yun, Weiliang Qiu, Feng Guo, Chunfang Huang, John Dominic Mancini, Kushagra Gupta, Maria E Laucho-Contreras, Zun Zar Chi Naing, Li Zhang, Mark A Perrella, Caroline A Owen, Edwin K Silverman, Xiaobo Zhou
ABSTRACT
Genetic variants in Hedgehog interacting protein (HHIP) have consistently been associated with the susceptibility to develop chronic obstructive pulmonary disease and pulmonary function levels, including the forced expiratory volume in 1 s (FEV1), in general population samples by genome-wide association studies. However, in vivo evidence connecting Hhip to age-related FEV1 decline and emphysema development is lacking. Herein, using Hhip heterozygous mice (Hhip(+/-)), we observed increased lung compliance and spontaneous emphysema in Hhip(+/-) mice starting at 10 mo of age. This increase was preceded by increases in oxidative stress levels in the lungs of Hhip(+/-) vs. Hhip(+/+) mice. To our knowledge, these results provide the first line of evidence that HHIP is involved in maintaining normal lung function and alveolar structures. Interestingly, antioxidant N-acetyl cysteine treatment in mice starting at age of 5 mo improved lung function and prevented emphysema development in Hhip(+/-) mice, suggesting that N-acetyl cysteine treatment limits the progression of age-related emphysema in Hhip(+/-) mice. Therefore, reduced lung function and age-related spontaneous emphysema development in Hhip(+/-) mice may be caused by increased oxidative stress levels in murine lungs as a result of haploinsufficiency of Hhip.
Sigma-Aldrich
Monoclonal Anti-HHIP antibody produced in mouse, clone 5D11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Millipore
MILLIPLEX® Mouse Cytokine/Chemokine Magnetic Bead Panel - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Millipore
MILLIPLEX® Mouse MMP Magnetic Bead Panel 3 - Immunology Multiplex Assay, MMP Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple MMP biomarkers in mouse cell culture samples.