Merck
  • Myxoid hepatocellular adenoma, a rare variant of hepatocellular adenoma with distinct imaging features: A case report with immunohistochemical and molecular analysis and literature review.
Myxoid hepatocellular adenoma, a rare variant of hepatocellular adenoma with distinct imaging features: A case report with immunohistochemical and molecular analysis and literature review.
Clinics and research in hepatology and gastroenterology (2020-07-06)
Nicolas De Vos, Joni Van der Meulen, Malaïka Van Der Linden, Kathleen Claes, Ann-Sophie Candaele, Aude Vanlander, Roberto Ivan Troisi, Hans Van Vlierberghe, Peter Smeets, Jo Van Dorpe, Anne Hoorens
ABSTRACT
Preoperative imaging and histopathology, immunohistochemistry and molecular analysis after resection of 2 hepatocellular adenomas (HCAs) (20 and 2cm) in a 53-year-old female patient were performed. On imaging, the large lesion resembled a myxoid HCA, while the small lesion resembled a more conventional HCA with a small myxoid/fluid area. On microscopy, the large lesion showed cords and nests of hepatocytes embedded in abundant myxoid matrix, while the small lesion resembled a conventional HCA with small foci of myxoid change and serosities; both consistent with a myxoid HCA. Immunophenotyping and molecular subtyping excluded inflammatory HCA, CTNNB1 mutated HCA and sonic hedgehog HCA, and was consistent with HNF1A mutated HCA. The myxoid change as well as the serosities may allow imaging diagnosis of myxoid HCA. As fluid vacuoles can also be present in ASS1+HCA, sonic hedgehog HCA has to be considered in the differential diagnosis.
Sigma-Aldrich
Anti-Arginase-1 Antibody, from chicken
Sigma-Aldrich
Anti-ASS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-PTGDS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution