Merck
  • CD93 regulates central nervous system inflammation in two mouse models of autoimmune encephalomyelitis.
CD93 regulates central nervous system inflammation in two mouse models of autoimmune encephalomyelitis.
Immunology (2018-06-21)
Mark R Griffiths, Marina Botto, Bryan Paul Morgan, James W Neal, Philippe Gasque
ABSTRACT
Microglia and non-professional immune cells (endothelial cells, neurons) participate in the recognition and removal of pathogens and tissue debris in the injured central nervous system through major pro-inflammatory processes. However, the mechanisms involved in regulating these responses remain ill-characterized. We herein show that CD93, also known as complement C1qRp/AA4 stem cell marker, has an important role in the regulation of inflammatory processes. The role of CD93 was evaluated in two models of neuroinflammation. We used the MOG-experimental autoimmune encephalomyelitis (EAE) model and the antibody-dependent EAE (ADEAE), which were induced in wild-type and CD93 knockout mice. We found that CD93 was highly expressed by neurons, endothelial cells and microglia (ramified >> amoeboid). Astrocytes and oligodendrocytes did not to express CD93. We further observed that CD93-deficient (CD93-/- ) mice presented a more robust brain and spinal cord inflammation in EAE and ADEAE. Encephalitis in CD93-/- was characterized by increased numbers of infiltrating M1 macrophages (CD11c+ CD206- ) and amoeboid microglia exhibiting a more activated phenotype (Tomato Lectinhigh Cox2high ). Damage to and leakage through the blood-brain barrier was increased in CD93-/- animals and was associated with a more robust neuronal injury when compared with wild-type EAE mice. We propose that CD93 is an important neuro-immune regulator to control central nervous system inflammation.
Sigma-Aldrich
Anti-DNA Antibody, single stranded specific, clone F7-26, clone F7-26, Chemicon®, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Nitric Oxide Synthase I Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Cyclooxygenase-2 Antibody, Chemicon®, from rabbit