Merck
  • VEGF-C, VEGF-A and related angiogenesis factors as biomarkers of allograft vasculopathy in cardiac transplant recipients.
VEGF-C, VEGF-A and related angiogenesis factors as biomarkers of allograft vasculopathy in cardiac transplant recipients.
The Journal of heart and lung transplantation : the official publication of the International Society for Heart Transplantation (2012-12-25)
Kevin P Daly, Michael E Seifert, Anil Chandraker, David Zurakowski, Anju Nohria, Michael M Givertz, S Ananth Karumanchi, David M Briscoe
ABSTRACT
Cardiac allograft vasculopathy (CAV), the major cause of late allograft loss after cardiac transplantation, results from donor-directed cellular and humoral alloimmune responses. Graft vascular endothelial cells (EC) are primary targets of these destructive responses, suggesting that factors associated with endothelial injury and repair could serve as biomarkers of CAV. Using a protein profiler array platform, we measured the levels of 55 angiogenesis-related proteins in sera from 33 adult heart transplant recipients, including 17 with angiographically documented CAV and 16 age- and gender-matched controls without CAV. All patients were >2 years after heart transplant. The study population was 75% male with a mean age of 62 ± 11 years. On average, patients were 12 ± 5 years after heart transplantation. We found that vascular endothelial growth factor (VEGF)-C, VEGF-A, angiopoietin-2, artemin, urokinase-type plasminogen activator and vasohibin were strongly associated with established CAV (all p < 0.01). Multivariable modeling identified VEGF-C, VEGF-A and platelet factor-4 (PF-4) as significant independent biomarkers of CAV. Furthermore, receiver-operating characteristic curve analysis demonstrated that the combination of all 3 molecules provided outstanding performance for the diagnosis of CAV (area under the curve [AUC] = 0.98; p < 0.001). Serum levels of VEGF-C, VEGF-A and PF-4 demonstrate strong associations with established CAV and, together with related angiogenesis factors, may serve as a reliable, non-invasive diagnostic test for CAV in cardiac transplant recipients.
Product Description
Millipore
MILLIPLEX® Human Angiogenesis/Growth Factor Magnetic Bead Panel - Cancer Multiplex Assay, Angiogenesie Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple angiogenic biomarkers in human serum, plasma and cell culture samples.