Merck
  • Impairment of chondrogenesis and microfibrillar network in Adamtsl2 deficiency.
Impairment of chondrogenesis and microfibrillar network in Adamtsl2 deficiency.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2018-10-12)
Laure Delhon, Clémentine Mahaut, Nicolas Goudin, Emilie Gaudas, Kevin Piquand, Wilfried Le Goff, Valérie Cormier-Daire, Carine Le Goff
ABSTRACT
Mutations in the a disintegrin and metalloproteinase with thrombospondin motif-like 2 ( ADAMTSL2) gene are responsible for the autosomal recessive form of geleophysic dysplasia, which is characterized by short stature, short extremities, and skeletal abnormalities. However, the exact function of ADAMTSL2 is unknown. To elucidate the role of this protein in skeletal development, we generated complementary knockout (KO) mouse models with either total or chondrocyte Adamtsl2 deficiency. We observed that the Adamtsl2 KO mice displayed skeletal abnormalities reminiscent of the human phenotype. Adamtsl2 deletion affected the growth plate formation with abnormal differentiation and proliferation of chondrocytes. In addition, a TGF-β signaling impairment in limbs lacking Adamtsl2 was demonstrated. Further investigations revealed that Adamtsl2 KO chondrocytes failed to establish a microfibrillar network composed by fibrillin1 and latent TGF-β binding protein 1 fibrils. Chondrocyte Adamtsl2 KO mice also exhibited dwarfism. These studies uncover the function of Adamtsl2 in the maintenance of the growth plate ECM by modulating the microfibrillar network.-Delhon, L., Mahaut, C., Goudin, N., Gaudas, E., Piquand, K., Le Goff, W., Cormier-Daire, V., Le Goff, C. Impairment of chondrogenesis and microfibrillar network in Adamtsl2 deficiency.
Sigma-Aldrich
Anti-Fibrillin-1 antibody produced in rabbit, affinity isolated antibody