Merck
  • Correction to: Multiparametric assessment of mitochondrial respiratory inhibition in HepG2 and RPTEC/TERT1 cells using a panel of mitochondrial targeting agrochemicals.
Correction to: Multiparametric assessment of mitochondrial respiratory inhibition in HepG2 and RPTEC/TERT1 cells using a panel of mitochondrial targeting agrochemicals.
Archives of toxicology (2020-07-29)
Wanda van der Stel, Giada Carta, Julie Eakins, Salihanur Darici, Johannes Delp, Anna Forsby, Susanne Hougaard Bennekou, Iain Gardner, Marcel Leist, Erik H J Danen, Paul Walker, Bob van de Water, Paul Jennings
ABSTRACT
In the original publication of the article.
Sigma-Aldrich
(−)-Deguelin, >98% (HPLC), powder