Merck
  • Adrenomedullin-CALCRL axis controls relapse-initiating drug tolerant acute myeloid leukemia cells.
Adrenomedullin-CALCRL axis controls relapse-initiating drug tolerant acute myeloid leukemia cells.
Nature communications (2021-01-20)
Clément Larrue, Nathan Guiraud, Pierre-Luc Mouchel, Marine Dubois, Thomas Farge, Mathilde Gotanègre, Claudie Bosc, Estelle Saland, Marie-Laure Nicolau-Travers, Marie Sabatier, Nizar Serhan, Ambrine Sahal, Emeline Boet, Sarah Mouche, Quentin Heydt, Nesrine Aroua, Lucille Stuani, Tony Kaoma, Linus Angenendt, Jan-Henrik Mikesch, Christoph Schliemann, François Vergez, Jérôme Tamburini, Christian Récher, Jean-Emmanuel Sarry
ABSTRACT
Drug tolerant/resistant leukemic stem cell (LSC) subpopulations may explain frequent relapses in acute myeloid leukemia (AML), suggesting that these relapse-initiating cells (RICs) persistent after chemotherapy represent bona fide targets to prevent drug resistance and relapse. We uncover that calcitonin receptor-like receptor (CALCRL) is expressed in RICs, and that the overexpression of CALCRL and/or of its ligand adrenomedullin (ADM), and not CGRP, correlates to adverse outcome in AML. CALCRL knockdown impairs leukemic growth, decreases LSC frequency, and sensitizes to cytarabine in patient-derived xenograft models. Mechanistically, the ADM-CALCRL axis drives cell cycle, DNA repair, and mitochondrial OxPHOS function of AML blasts dependent on E2F1 and BCL2. Finally, CALCRL depletion reduces LSC frequency of RICs post-chemotherapy in vivo. In summary, our data highlight a critical role of ADM-CALCRL in post-chemotherapy persistence of these cells, and disclose a promising therapeutic target to prevent relapse in AML.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
MISSION® esiRNA, targeting human CALCRL
Sigma-Aldrich
Hexadimethrine bromide, ≥95%